2-Phenylethyl isovalerate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Phenethyl 3-methylbutanoate, Phenethyl Isovalerate, Isovaleric Acid 2-Phenylethyl Ester, Isovaleric Acid Phenethyl Ester

P2009-25ML P2009-500ML
TCIAP2009-25MLEA 215 SEK
TCIAP2009-25ML TCIAP2009-500ML
2-Phenylethyl isovalerate ≥98.0%
2-Phenylethyl isovalerate
Formel: C₁₃H₁₈O₂
MW: 206,29 g/mol
Kokpunkt: 268°C
Flampunkt: 100°C
MDL Number: MFCD00044322
CAS nummer: 140-26-1
EINECS: 205-406-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d39dfefa423b0a9535e773e5485d3019773d43a&vid=1d39dfefa423b0a9535e773e5485d3019773d43a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR