Pemirolast potassium, hydrate ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Potassium 5-(9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl)tetrazol-2-ide, 9-Methyl-3-(1H-tetrazol-5-yl)-4H-pyrido[1,2-a]pyrimidin-4-one potassium salt
P1995-5G P1995-1G
TCIAP1995-5GEA 3275 SEK
TCIAP1995-5G TCIAP1995-1G
Pemirolast potassium, hydrate ≥98.0% (by HPLC, titration analysis)
Pemirolast potassium
Formel: C₁₀H₇KN₆O
MW: 266,305 g/mol
Smältpunkt: 317°C
Storage Temperature: Kylskåp
MDL Number: MFCD01690051
CAS nummer: 100299-08-9
Merck Index: 14,07078
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 3.0 to 7.0 %