MOPS, Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
475898-1 475898-100 475898-500
475898-1EA 13050 SEK
475898-1 475898-100 475898-500
MOPS, Millipore®
MOPS
Formel: C₇H₁₅NO₄S
MW: 209,27 g/mol
Smältpunkt: > 250 °C
Flampunkt: 110 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00006183
CAS nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288