Perphenazine ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazineethanol, 2-Chloro-10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazine
P1970-5G
P1970-25G
TCIAP1970-5GEA
1125
SEK
TCIAP1970-5G
TCIAP1970-25G
Perphenazine ≥97.0% (by HPLC, titration analysis)
Perphenazine
Formel:
C₂₁H₂₆ClN₃OS MW: 403,97 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00056798 CAS nummer: 58-39-9 EINECS: 200-381-5 Merck Index: 14,07183 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...