Pretilachlor ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Chloro-2',6'-diethyl-N-(2-propoxyethyl)acetanilide
P1967-25G P1967-5G
TCIAP1967-25GEA 3125 SEK
TCIAP1967-25G TCIAP1967-5G
Pretilachlor ≥97.0%
Pretilachlor
Formel: C₁₇H₂₆ClNO₂
MW: 311,85 g/mol
Kokpunkt: 129°C
Storage Temperature: Kylskåp
MDL Number: MFCD00161476
CAS nummer: 51218-49-6
UN: 3082
ADR: 9,III
Merck Index: 14,07740
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %