Pendimethalin 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 3,4-Dimethyl-2,6-dinitro-N-(3-pentyl)aniline

P1964-5G P1964-25G P1964-1G
TCIAP1964-5GEA 1125 SEK
TCIAP1964-5G TCIAP1964-25G TCIAP1964-1G
Pendimethalin 98,0 GC_ANALYSMETOD
Pendimethalin
Formel: C₁₃H₁₉N₃O₄
MW: 281.31 g/mol
MDL Number: MFCD00055332
CAS nummer: 40487-42-1
EINECS: 254-938-2
UN: 3077
ADR: 9 III
Merck Index: 14,07084
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e59af7e91ddda3dac76fbbf6e4f461ed7e4b883d&vid=e59af7e91ddda3dac76fbbf6e4f461ed7e4b883d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR