3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥98.0%
Leverantör: TCI EUROPE
Varning
P1963-1G P1963-200MG
TCIAP1963-1GEA 4800 SEK
TCIAP1963-1G TCIAP1963-200MG
3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥98.0%
3-Phenyl-3-(trifluoromethyl)-3H-diazirine
Formel: C₈H₅F₃N₂
MW: 186,14 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22041649
CAS nummer: 73899-14-6
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%