(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol

P1934-25G P1934-5G
TCIAP1934-25GEA 1200 SEK
TCIAP1934-25G TCIAP1934-5G
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol ≥98.0%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Formel: C₁₀H₁₈O₂
MW: 170,25 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00077851
CAS nummer: 18680-27-8

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Specific rotation [a]20/D +8.0 to +11.0 deg (C=6.5, toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR