Prostaglandin E2 ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
PGE₂
P1884-1MG
P1884-10MG
TCIAP1884-1MGEA
525
SEK
TCIAP1884-1MG
TCIAP1884-10MG
Prostaglandin E2 ≥98.0%
Prostaglandin E2
Formel:
C₂₀H₃₂O₅ MW: 352,47 g/mol Smältpunkt: 66°C Storage Temperature: Frys |
MDL Number:
MFCD00077861 CAS nummer: 363-24-6 EINECS: 206-656-6 Merck Index: 14,07877 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...