Piceid ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Resveratrol 3-Glucoside, (E)-Piceid, 4',5-Dihydroxystilben-3-yl beta-D-Glucopyranoside, Polydatin, trans-Resveratrol-3-O-β-D-Glucopyranoside, trans-Polydatin, (E)-Polydatin, 3,4,5-Trihydroxystilbene-3-beta-monoglucoside, 3,4,5-Tsg, (2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

P1878-1G P1878-5G
TCIAP1878-1GEA 430 SEK
TCIAP1878-1G TCIAP1878-5G
Piceid ≥95.0%
Piceid
Formel: C₂₀H₂₂O₈
MW: 390,39 g/mol
Smältpunkt: 226°C
Storage Temperature: Kylskåp
MDL Number: MFCD00210592
CAS nummer: 27208-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b239458f770526c784d72c14c8b582633d1e5f24&vid=b239458f770526c784d72c14c8b582633d1e5f24&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Specific rotation [a]20/D -65.0 to -70.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR