Pentabromobenzyl acrylate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Acrylic acid Pentabromobenzyl ester

P1877-1G P1877-5G
TCIAP1877-1GEA 885 SEK
TCIAP1877-1G TCIAP1877-5G
Pentabromobenzyl acrylate ≥98.0%
Pentabromobenzyl acrylate
Formel: C₁₀H₅Br₅O₂
MW: 556,67 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00804467
CAS nummer: 59447-55-1
EINECS: 261-767-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b450d64cb9634404184ff594a28972d43a0fb90e&vid=b450d64cb9634404184ff594a28972d43a0fb90e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR