Pravastatin sodium ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
(3R,5R)-3,5-Dihydroxy-7-((1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-(((S)-2-methylbutanoyl)oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl)heptanoic acid, sodium salt
P1796-100MG
P1796-500MG
TCIAP1796-100MGEA
1300
SEK
TCIAP1796-100MG
TCIAP1796-500MG
Pravastatin sodium ≥98.0% (by HPLC, titration analysis)
Pravastatin sodium
Formel:
C₂₃H₃₅NaO₇ MW: 446,52 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00887601 CAS nummer: 81131-70-6 Merck Index: 14,07714 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +153 to +159 deg (C=0.5, H2O) (calcd.on anh.substance) |
Water | max. 4.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...