Piperitone (mixture of enantiomers) (predominantly (R)-(-)-form) ≥94.0%

Leverantör: TCI
Varning

P1783-25G P1783-100G
TCIAP1783-25GEA 495 SEK
TCIAP1783-25G TCIAP1783-100G
Piperitone (mixture of enantiomers) (predominantly (R)-(-)-form) ≥94.0%
Piperitone (mixture of enantiomers)
Formel: C₁₀H₁₆O
Kokpunkt: 235°C
Smältpunkt: –29°C
CAS nummer: 89-81-6
EINECS: 224-957-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1030a6ac3c43053387e079e70885813d7a07219e&vid=19514ab2b4abd380b26ce8b73780d1888886e1c1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 94.0 %
Optical purity (LC) 70.0 to 90.0 ee%
Specific rotation [a]20/D -45.0 to -60.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR