1,2,2,3-Propanetetracarbonitrile ≥96.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,2,2,3-Tetracyanopropane

P1751-1G P1751-5G
TCIAP1751-1GEA 1600 SEK
TCIAP1751-1G TCIAP1751-5G
1,2,2,3-Propanetetracarbonitrile ≥96.0%
1,2,2,3-Propanetetracarbonitrile
Formel: C₇H₄N₄
MW: 144,14 g/mol
CAS nummer: 1274904-48-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70940d05808ce007ec12f41d329731715ffdd2c2&vid=70940d05808ce007ec12f41d329731715ffdd2c2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR