MOPS ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
A12914.14 A12914.22 A12914.36
A12914.14EA 740 SEK
A12914.14 A12914.22 A12914.36
MOPS ≥99%
MOPS
Purity: 99% CAS No.: 1132-61-2 Molecular Formula: C7H15NO4S Formula Weight: 209.27 Flash Point: 110°(230°F) Physical Form: Powder Melting Point: 277-280° dec. Beilstein Reference: 1106776 Merck Reference: 14,6265 MDL No.: MFCD00006183
Good's buffer
Formel: C₇H₁₅NO₄S
MW: 209,27 g/mol
Smältpunkt: 277…280 °C (dec.)
Flampunkt: 110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006183
CAS nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 14,6265