6,13-Pentacenedione ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 6,13-Pentacenequinone
P1685-25G P1685-5G
TCIAP1685-25GEA 5800 SEK
TCIAP1685-25G TCIAP1685-5G
6,13-Pentacenedione ≥95.0%
6,13-Pentacenedione
Formel: C₂₂H₁₂O₂
MW: 308,34 g/mol
Smältpunkt: 394°C
MDL Number: MFCD00003709
CAS nummer: 3029-32-1
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%