Pentamethyldisiloxane ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,1,1,3,3-Pentamethyldisiloxane

P1535-5ML P1535-25ML
TCIAP1535-5MLEA 455 SEK
TCIAP1535-5ML TCIAP1535-25ML
Pentamethyldisiloxane ≥95.0%
Pentamethyldisiloxane
Formel: C₅H₁₆OSi₂
MW: 148,35 g/mol
Kokpunkt: 86°C
Flampunkt: -19°C
MDL Number: MFCD00053658
CAS nummer: 1438-82-0
EINECS: 215-873-5
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54c87d72c76e1559d185c5f1b1eab7568c13ab90&vid=54c87d72c76e1559d185c5f1b1eab7568c13ab90&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR