1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate ≥97.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Methacrylic Acid 1,2,2,6,6-Pentamethyl-4-piperidyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P1513-25G P1513-500G
TCIAP1513-25GEA 685 SEK
TCIAP1513-25G TCIAP1513-500G
1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate ≥97.0% stabilised
1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate
Formel: C₁₄H₂₅NO₂
MW: 239,36 g/mol
Flampunkt: 112°C
Storage Temperature: Kylskåp
CAS nummer: 68548-08-3
EINECS: 271-354-3

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR