N-Phenyl-9-anthramine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: N-Phenylanthracen-9-amine, 9-Anilinoanthracene

P1496-1G P1496-5G
TCIAP1496-1GEA 805 SEK
TCIAP1496-1G TCIAP1496-5G
N-Phenyl-9-anthramine ≥98.0%
N-Phenyl-9-anthramine
Formel: C₂₀H₁₅N
MW: 269,35 g/mol
Smältpunkt: 207°C
MDL Number: MFCD03093237
CAS nummer: 15424-38-1

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 210.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR