(R)-Propylene carbonate ≥98.0%
Leverantör: TCI
|
Varning
|
P1485-25G
P1485-5G
TCIAP1485-25GEA
2225
SEK
TCIAP1485-25G
TCIAP1485-5G
(R)-Propylene carbonate ≥98.0%
(R)-Propylene carbonate
Formel:
C₄H₆O₃ Flampunkt: 132°C |
MDL Number:
MFCD00798265 CAS nummer: 16606-55-6 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...