Potassium bis(1-pyrazolyl)borohydride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Potassium Dihydrobis(1-pyrazolyl)borate

P1439-1G P1439-5G
TCIAP1439-1GEA 845 SEK
TCIAP1439-1G TCIAP1439-5G
Potassium bis(1-pyrazolyl)borohydride ≥98.0%
Potassium bis(1-pyrazolyl)borohydride
Formel: C₆H₈BKN₄
MW: 186,07 g/mol
MDL Number: MFCD00058739
CAS nummer: 18583-59-0
UN: 2813
ADR: 4.3,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR