Diethyl-1,4-phenylenediacetate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Diethyl 2,2-(1,4-phenylene)diacetate, 1,4-Phenylenediacetic Acid Diethyl Ester

P1434-25G P1434-5G
TCIAP1434-25GEA 4100 SEK
TCIAP1434-25G TCIAP1434-5G
Diethyl-1,4-phenylenediacetate ≥98.0%
Diethyl-1,4-phenylenediacetate
Formel: C₁₄H₁₈O₄
MW: 250,29 g/mol
Kokpunkt: 140°C
MDL Number: MFCD00032366
CAS nummer: 36076-26-3

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR