Methyl Phenylsulphonylacetate ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Phenylsulphonylacetic Acid Methyl Ester

P1353-25G P1353-5G
TCIAP1353-25GEA 1600 SEK
TCIAP1353-25G TCIAP1353-5G
Methyl Phenylsulphonylacetate ≥95.0%
Methyl Phenylsulphonylacetate
Formel: C₉H₁₀O₄S
MW: 214,24 g/mol
Flampunkt: 78°C
MDL Number: MFCD00007555
CAS nummer: 34097-60-4

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid to slightly cloudy liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR