Phenyl-β-D-galactopyranoside ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Phenyl b-D-galactoside

P1326-1G P1326-5G
TCIAP1326-1GEA 710 SEK
TCIAP1326-1G TCIAP1326-5G
Phenyl-β-D-galactopyranoside ≥98.0%
Phenyl-beta-D-galactopyranoside
Formel: C₁₂H₁₆O₆
MW: 256,25 g/mol
MDL Number: MFCD00063258
CAS nummer: 2818-58-8
EINECS: 220-573-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 146.0 to 149.0 deg-C
Specific rotation [a]20/D -40.0 to -44.0 deg (C=2.3, H2O)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR