Phenyl vinyl sulphide ≥98.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: (Phenylthio)ethylene

Stabilization: med 4-tert-Butylpyrokatekol

P1287-25G P1287-5G
TCIAP1287-25GEA 3825 SEK
TCIAP1287-25G TCIAP1287-5G
Phenyl vinyl sulphide ≥98.0% stabilised
Phenyl vinyl sulphide
Formel: C₈H₈S
MW: 136,21 g/mol
Kokpunkt: 78°C
Flampunkt: 46°C
MDL Number: MFCD00009646
CAS nummer: 1822-73-7
EINECS: 217-351-2
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR