Diethyl (phthalimidomethyl)phosphonate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Diethyl ((1,3-dioxoisoindolin-2-yl)methyl)phosphonate, (Phthalimidomethyl)phosphonic Acid Diethyl Ester

P1193-25G P1193-5G
TCIAP1193-25GEA 2600 SEK
TCIAP1193-25G TCIAP1193-5G
Diethyl (phthalimidomethyl)phosphonate ≥98.0%
Diethyl (phthalimidomethyl)phosphonate
Formel: C₁₃H₁₆NO₅P
MW: 297,25 g/mol
MDL Number: MFCD00134313
CAS nummer: 33512-26-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR