O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
P1190-1G
P1190-100MG
P1190-5G
TCIAP1190-1GEA
4000
SEK
TCIAP1190-1G
TCIAP1190-100MG
TCIAP1190-5G
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime ≥98.0%
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime
Formel:
C₈H₄F₅NO |
MDL Number:
MFCD00191477 CAS nummer: 86356-73-2 |
Specifikation testresultat
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 23.0 to 25.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...