1-[4-(2-Phenylethyl)benzyl]naphthalene ≥98.0%

Leverantör: TCI

P1181-1G P1181-25G
TCIAP1181-1GEA 765 SEK
TCIAP1181-1G TCIAP1181-25G
1-[4-(2-Phenylethyl)benzyl]naphthalene ≥98.0%
1-[4-(2-Phenylethyl)benzyl]naphthalene
Formel: C₂₅H₂₂
MDL Number: MFCD00191572
CAS nummer: 127833-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=701a0faf728ea1911b7a2fa0a81595019e8bf903&vid=701a0faf728ea1911b7a2fa0a81595019e8bf903&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR