Pyromellitic diimide ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Benzene-1,2,4,5-tetracarboxylic Diimide

P1153-25G P1153-5G
TCIAP1153-25GEA 2275 SEK
TCIAP1153-25G TCIAP1153-5G
Pyromellitic diimide ≥97.0%
Pyromellitic diimide
Formel: C₁₀H₄N₂O₄
MW: 216,15 g/mol
MDL Number: MFCD00005004
CAS nummer: 2550-73-4
EINECS: 219-852-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR