(R)-3-Hydroxypyrrolidine hydrochloride ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(R)-3-Pyrrolidinol hydrochloride, (3R)-pyrrolidin-3-ol hydrochloride, (R)-3-Hydroxypyrrolidine HCl
P1140-25G
P1140-5G
TCIAP1140-25GEA
3500
SEK
TCIAP1140-25G
TCIAP1140-5G
(R)-3-Hydroxypyrrolidine hydrochloride ≥98.0%
(R)-3-Hydroxypyrrolidine hydrochloride
Formel:
C₄H₁₀ClNO MW: 123,58 g/mol |
MDL Number:
MFCD00191570 CAS nummer: 104706-47-0 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Specific rotation [a]20/D | -7.5 to -8.0 deg (C=3.5, Methanol) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...