3-Propoxypropylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Amino-3-propoxypropane

P1095-25ML P1095-5ML
TCIAP1095-25MLEA 4675 SEK
TCIAP1095-25ML TCIAP1095-5ML
3-Propoxypropylamine ≥98.0% (by GC, titration analysis)
3-Propoxypropylamine
Formel: C₆H₁₅NO
MW: 117,19 g/mol
Kokpunkt: 152°C
MDL Number: MFCD00129001
CAS nummer: 16728-59-9
EINECS: 240-792-7
UN: 2733
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR