[Hydroxy(tosyloxy)iodo]benzene ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Koser Reagent, Phenyliodosohydroxy Tosylate
P1015-25G
P1015-5G
TCIAP1015-25GEA
4025
SEK
TCIAP1015-25G
TCIAP1015-5G
[Hydroxy(tosyloxy)iodo]benzene ≥97.0%
[Hydroxy(tosyloxy)iodo]benzene
Formel:
C₁₃H₁₃IO₄S |
MDL Number:
MFCD00011547 CAS nummer: 27126-76-7 Merck Index: 14,05321 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Purity (Iodometric Titration) | min. 97.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...