Didecyl phosphate ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Phosphoric Acid Didecyl Ester

P1011-1G P1011-100MG P1011-5G
TCIAP1011-1GEA 2450 SEK
TCIAP1011-1G TCIAP1011-100MG TCIAP1011-5G
Didecyl phosphate ≥95.0%
Didecyl phosphate
Formel: C₂₀H₄₃O₄P
MW: 378,53 g/mol
MDL Number: MFCD00015062
CAS nummer: 7795-87-1
EINECS: 232-248-2

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 46.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR