Pyridinium-3-nitrobenzenesulphonate ≥98.0%

Leverantör: TCI EUROPE
Varning

P0941-25G P0941-5G
TCIAP0941-25GEA 1275 SEK
TCIAP0941-25G TCIAP0941-5G
Pyridinium-3-nitrobenzenesulphonate ≥98.0%
Pyridinium-3-nitrobenzenesulphonate
Formel: C₁₁H₁₀N₂O₅S
MW: 282,27 g/mol
MDL Number: MFCD00060234
CAS nummer: 84752-61-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 122.0 to 125.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR