Bis(dimethylamino)phosphoryl chloride ≥93.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Phosphoric Acid Bis(dimethylamido) Chloride, N,N,N',N'-Tetramethylphosphorodiamidic Chloride, Bis(dimethylamino)phosphorochloridate, Bis(dimethylamido)phosphoric chloride

P0937-25G P0937-5G
TCIAP0937-25GEA 2150 SEK
TCIAP0937-25G TCIAP0937-5G
Bis(dimethylamino)phosphoryl chloride ≥93.0%
Bis(dimethylamino)phosphoryl chloride
Formel: C₄H₁₂ClN₂OP
MW: 170,58 g/mol
Kokpunkt: 110°C
MDL Number: MFCD00008302
CAS nummer: 1605-65-8
EINECS: 216-517-1
UN: 3265
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR