Pentaerythritol tetrakis(mercaptoacetate) ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Pentaerythritol Tetrathioglycolate
P0888-25G
P0888-500G
TCIAP0888-25GEA
215
SEK
TCIAP0888-25G
TCIAP0888-500G
Pentaerythritol tetrakis(mercaptoacetate) ≥95.0%
Pentaerythritol tetrakis(mercaptoacetate)
Formel:
C₁₃H₂₀O₈S₄ MW: 432,54 g/mol Flampunkt: 248°C |
MDL Number:
MFCD00022080 CAS nummer: 10193-99-4 EINECS: 233-482-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (Iodometric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...