Ethyl pentadecanoate ≥97.0%
Leverantör: TCI
Synonyms:
Pentadecanoic Acid Ethyl Ester
P0868-25ML
P0868-5ML
TCIAP0868-25MLEA
3025
SEK
TCIAP0868-25ML
TCIAP0868-5ML
Ethyl pentadecanoate ≥97.0%
Ethyl pentadecanoate
Formel:
C₁₇H₃₄O₂ Kokpunkt: 158°C |
MDL Number:
MFCD00042872 CAS nummer: 41114-00-5 EINECS: 255-223-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...