(-)-Perillaldehyde ≥90.0%

Leverantör: TCI EUROPE
Varning

Synonyms: L(-)-Perillaldehyde, L-Perillaldehyde, (-)-4-Isopropenyl-1-cyclohexene-1-carboxaldehyde, (S)-4-(Prop-1-en-2-yl)cyclohex-1-ene-1-carbaldehyde

P0866-25ML P0866-5ML
TCIAP0866-25MLEA 1100 SEK
TCIAP0866-25ML TCIAP0866-5ML
(-)-Perillaldehyde ≥90.0%
(-)-Perillaldehyde
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokpunkt: 98°C
Flampunkt: 95°C
MDL Number: MFCD00001543
CAS nummer: 18031-40-8
Merck Index: 14,07167
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a423cef615537f0476002c4382b90337179a92fc&vid=a423cef615537f0476002c4382b90337179a92fc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 90.0 %
Specific rotation [a]20/D -135 to -124 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR