Phenyl vinyl sulphoxide ≥95.0%

Leverantör: TCI EUROPE

Synonyms: (Vinylsulphinyl)benzene

P0863-25G P0863-5G
TCIAP0863-25GEA 5175 SEK
TCIAP0863-25G TCIAP0863-5G
Phenyl vinyl sulphoxide ≥95.0%
Phenyl vinyl sulphoxide
Formel: C₈H₈OS
MW: 152,21 g/mol
Kokpunkt: 110°C
MDL Number: MFCD00002086
CAS nummer: 20451-53-0
EINECS: 243-831-6

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.1580 to 1.1640
Refractive index n20/D 1.5840 to 1.5880

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR