Pentaerythritol tetraacetate ≥98.0%
Leverantör: TCI EUROPE
P0757-25G
P0757-500G
TCIAP0757-25GEA
455
SEK
TCIAP0757-25G
TCIAP0757-500G
Pentaerythritol tetraacetate ≥98.0%
Pentaerythritol tetraacetate
Formel:
C₁₃H₂₀O₈ Kokpunkt: 325°C |
MDL Number:
MFCD00026206 CAS nummer: 597-71-7 EINECS: 209-907-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 83.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...