Phenylthiohydantoin-proline ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: PTH-proline, Phenylthiohydantoin-proline, 2-Phenyl-3-thioxohexahydro-1H-pyrrolo[1,2-c]imidazol-1-one

P0729-1G P0729-100MG
TCIAP0729-1GEA 4275 SEK
TCIAP0729-1G TCIAP0729-100MG
Phenylthiohydantoin-proline ≥98.0% (by HPLC, total nitrogen)
Phenylthiohydantoin-proline
Formel: C₁₂H₁₂N₂OS
MW: 232,30 g/mol
MDL Number: MFCD00056895
CAS nummer: 4333-21-5
EINECS: 224-374-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 177.0 to 181.0 deg-C
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR