Polyethylene glycol monooleyl ether n=∼7

Leverantör: TCI EUROPE
Varning

P0713-25G P0713-500G
TCIAP0713-25GEA 215 SEK
TCIAP0713-25G TCIAP0713-500G
Polyethylene glycol monooleyl ether n=∼7
Polyethylene glycol monooleyl ether
Formel: HO(CH₂CH₂O)nC₁₈H₃₅
MDL Number: MFCD00074369
CAS nummer: 9004-98-2
EINECS: 500-016-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23314cb6e6db35b34d785e85e51a4db479cc939e&vid=c203da7bcef0cef44f67808b768152ed0ee7b99b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

The average value of n 6.0 to 8.5
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR