Vinyl pivalate ≥99.0% stabilised
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Vinyl Trimethylacetate, Trimethylacetic Acid Vinyl Ester, Pivalic Acid Vinyl Ester
Stabilization:
med hydrokinon
P0668-25ML
P0668-500ML
TCIAP0668-25MLEA
215
SEK
TCIAP0668-25ML
TCIAP0668-500ML
Vinyl pivalate ≥99.0% stabilised
Vinyl pivalate
Formel:
C₇H₁₂O₂ MW: 128,17 g/mol Kokpunkt: 112°C Flampunkt: 15°C |
MDL Number:
MFCD00059211 CAS nummer: 3377-92-2 EINECS: 222-175-4 UN: 3272 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...