8-Hydroxy-7-propylquinoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
7-Propyloxine, 7-Propyl-8-quinolinol
P0531-1G
P0531-5G
TCIAP0531-1GEA
1850
SEK
TCIAP0531-1G
TCIAP0531-5G
8-Hydroxy-7-propylquinoline ≥98.0% (by GC, titration analysis)
8-Hydroxy-7-propylquinoline
Formel:
C₁₂H₁₃NO MW: 187,24 g/mol Kokpunkt: 149°C Smältpunkt: 0°C |
MDL Number:
MFCD00059755 CAS nummer: 58327-60-9 |
Specifikation testresultat
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...