Polyvinylpyrrolidone, M.W. ∼ 10000 K 15

Leverantör: TCI EUROPE

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P0471-25G P0471-500G P0471-100G
TCIAP0471-25GEA 270 SEK
TCIAP0471-25G TCIAP0471-500G TCIAP0471-100G
Polyvinylpyrrolidone, M.W. ∼ 10000 K 15
Polyvinylpyrrolidone
Formel: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS nummer: 9003-39-8
Merck Index: 14,07697

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Total Nitrogen 12.0 to 12.8 w/w% (calcd.on anh.substance)
K value 12.0 to 18.0
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR