Phytic acid ∼50% ∼1.1 mol/L i vatten
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Inositol hexaphosphoric acid
P0409-25G
P0409-500G
TCIAP0409-25GEA
245
SEK
TCIAP0409-25G
TCIAP0409-500G
Phytic acid ∼50% ∼1.1 mol/L i vatten
Phytic acid
Formel:
C₆H₁₈O₂₄P₆ MW: 660,03 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00082309 CAS nummer: 83-86-3 EINECS: 201-506-6 Merck Index: 14,07387 |
Specifikation testresultat
Concentration (Neutralization titration) | 47.0 to 54.0 w/w% |
Binding phosphorous | 13.5 to 14.6 % |
Free phosphorous | max. 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...