Triamyl phosphate ≥98.0%

Leverantör: TCI

Synonyms: Phosphoric Acid Triamyl Ester, Tripentyl Phosphate, Tri-n-amylphosphate

P0265-25ML P0265-5ML
TCIAP0265-25MLEA 2625 SEK
TCIAP0265-25ML TCIAP0265-5ML
Triamyl phosphate ≥98.0%
Triamyl phosphate
Formel: C₁₅H₃₃O₄P
MW: 308.40 g/mol
Kokpunkt: 158°C
MDL Number: MFCD00015348
CAS nummer: 2528-38-3
EINECS: 219-773-2

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR