Potassium Phenyl sulphate ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Sulphuric acid phenyl ester potassium salt
P0232-1G P0232-5G
TCIAP0232-1GEA 1550 SEK
TCIAP0232-1G TCIAP0232-5G
Potassium Phenyl sulphate ≥98.0% (by HPLC, titration analysis)
Potassium Phenyl sulphate
Formel: C₆H₅KO₄S
MW: 212,26 g/mol
Smältpunkt: 160°C
Storage Temperature: Frys
CAS nummer: 1733-88-6
EINECS: 217-071-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edc632843cc35d358d71290bea744e016f2452c2&vid=edc632843cc35d358d71290bea744e016f2452c2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %