Phenyl phosphate disodium salt ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Phosphoric acid phenyl ester disodium salt, Phosphoric acid monophenyl ester disodium salt

P0208-25G P0208-500G
TCIAP0208-25GEA 815 SEK
TCIAP0208-25G TCIAP0208-500G
Phenyl phosphate disodium salt ≥98.0%
Phenyl phosphate disodium salt
Formel: C₆H₅Na₂O₄P·xH₂O
MW: 218,06 g/mol (anhy)
MDL Number: MFCD00002133
CAS nummer: 3279-54-7
EINECS: 221-917-4
Merck Index: 14,03357

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 7.0 to 16.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR