Parabanic acid ≥98.0%
Leverantör: TCI
|
Varning
|
P0195-1G
P0195-25G
TCIAP0195-1GEA
445
SEK
TCIAP0195-1G
TCIAP0195-25G
Parabanic acid ≥98.0%
Parabanic acid
Formel:
C₃H₂N₂O₃ |
MDL Number:
MFCD00014493 CAS nummer: 120-89-8 EINECS: 204-434-3 Merck Index: 14,07022 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...